Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Fscn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107072.1 | NP_001100542.1 | 1 | 748 | ||
XM_008768476.1 | XP_008766698.1 | 1 | 5905 |