Search
Search

Rabif
RGD1563962| Gene Symbol: Rabif | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007678.2 | NP_001007679.1 | 2 | 257 | ||
| GenBank mRNA | BC161807.1 | - | 2 | 265 | 9036685 | |
| CF977877.1 | - | 2 | 281 | |||
| Gene Symbol: RGD1563962 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003749301.3 | XP_003749349.1 | 2 | 251 | ||