Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Cxcl11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182952.2 | NP_891997.1 | 2, 3 | 251 | ||
GenBank mRNA | AF537209.1 | - | 2, 3 | 251 | ||
AY340181.1 | - | 2, 3 | 230 | |||
BC168725.1 | - | 2, 3 | 259 | 6919932 |