Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Samd4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107254.1 | NP_001100724.1 | 2 | 634 | ||
XM_008770515.1 | XP_008768737.1 | 2 | 1472 | |||
XM_008770516.1 | XP_008768738.1 | 2 | 1471 | |||
GenBank mRNA | BC168894.1 | - | 2 | 616 | 7453870 |