Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other LOC100911492 SIRNA Assays ›
          Gene Symbol:
          LOC100911492 Otx2
          Gene Name:
          homeobox protein OTX2-like
          orthodenticle homeobox 2
          Gene Aliases:
          -
          -
          Chromosome Location:
          Chr.15: 25516697 - 25526560 on Build Rnor_6.0
          Chromosome Location:
          Chr.15: 25500037 - 25510602 on Build Rnor_6.0
          UniGene ID:
          -
          Rn.35222
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          -
          siRNA ID s157077

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price 5,354.00
          Your Price
          Online offer:
          5354.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: LOC100911492
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq XM_003751455.3 XP_003751503.1 5 2549
          XM_006251835.2 XP_006251897.1 5 2578
          XM_006251836.2 XP_006251898.1 6 1546
          XM_006251838.2 XP_006251900.1 5 1383
          XM_006251839.2 XP_006251901.1 5 1326
          XM_008770615.1 XP_008768837.1 8 2845
          XM_008770616.1 XP_008768838.1 6 1491
          XM_008770617.1 XP_008768839.1 6 2393
          XM_008770618.1 XP_008768840.1 5 1480
          XM_008770619.1 XP_008768841.1 8 2788
          XM_008770620.1 XP_008768842.1 6 1552
          XM_008770621.1 XP_008768843.1 6 1538
          XM_008770622.1 XP_008768844.1 6 1434
          XM_008770623.1 XP_008768845.1 5 2759
          XM_008770624.1 XP_008768846.1 6 2645
          Gene Symbol: Otx2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001100566.1 NP_001094036.1 1267
          XM_006251808.2 XP_006251870.1 2578
          XM_006251809.2 XP_006251871.1 2549
          XM_006251810.2 XP_006251872.1 1061
          XM_008770523.1 XP_008768745.1 2205
          XM_008770524.1 XP_008768746.1 1235
          XM_008770525.1 XP_008768747.1 2393
          XM_008770526.1 XP_008768748.1 1480
          XM_008770527.1 XP_008768749.1 1175
          XM_008770528.1 XP_008768750.1 2759
          XM_008770529.1 XP_008768751.1 2645
          XM_008770530.1 XP_008768752.1 1123

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Human: NM_001270523, NM_001270524, NM_021728, NM_172337, NM_001270525, and others.
          This siRNA also targets the following transcripts in Mouse: NM_001286481, NM_001286483, NM_144841, NM_001286482, AK087527, and others.

          Panther Classification:

          Molecular Function -

          homeodomain transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          cell fate specification
          regulation of transcription, DNA-templated
          regulation of transcription from RNA polymerase II promoter
          protein complex assembly
          multicellular organism development
          axon guidance
          central nervous system development
          endoderm development
          organ morphogenesis
          anterior/posterior pattern specification
          dorsal/ventral pattern formation
          metencephalon development
          cell differentiation
          neuron differentiation
          forebrain development
          midbrain development
          somite rostral/caudal axis specification
          positive regulation of embryonic development
          inner ear morphogenesis
          eye photoreceptor cell fate commitment
          cell fate commitment
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          neuron fate commitment
          neuron fate determination
          diencephalon morphogenesis
          anatomical structure development
          primitive streak formation
          positive regulation of gastrulation
          RNA polymerase II core promoter proximal region sequence-specific DNA binding
          transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
          transcription factor activity, sequence-specific DNA binding
          sequence-specific DNA binding
          protein heterodimerization activity
          DNA binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-55658555d6-7l2qm:80/100.66.75.127:80.
          git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.41.0-Offline