Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Zc3h13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170471.1 | NP_001163942.1 | 14 | 3200 | ||
XM_006252316.2 | XP_006252378.1 | 14 | 3256 | |||
XM_006252317.2 | XP_006252379.1 | 14 | 3260 |