Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: B3gnt8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107492.1 | NP_001100962.1 | 3 | 1119 | ||
XM_006228433.2 | XP_006228495.1 | 2 | 2517 | |||
XM_006228434.2 | XP_006228496.1 | 2 | 2559 | |||
XM_006228436.2 | XP_006228498.1 | 2 | 2205 |