Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Trim5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014023.1 | NP_001014045.1 | 2 | 228 | ||
XM_006229912.2 | XP_006229974.1 | 2 | 714 | |||
GenBank mRNA | BC078926.1 | - | 2 | 228 | 7106563 |