Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PLA2G6 SIRNA Assays ›
          Gene Symbol:
          PLA2G6
          Gene Name:
          phospholipase A2 group VI
          Gene Aliases:
          CaI-PLA2, GVI, INAD1, IPLA2-VIA, NBIA2, NBIA2A, NBIA2B, PARK14, PLA2, PNPLA9, iPLA2, iPLA2beta
          Chromosome Location:
          Chr.22: 38111495 - 38192099 on Build GRCh38
          UniGene ID:
          Hs.170479
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s15941

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: PLA2G6
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001004426.1 NP_001004426.1 11 1598
          NM_001199562.1 NP_001186491.1 11 1553
          NM_003560.2 NP_003551.2 12 1760
          XM_005261764.2 XP_005261821.1 12 1735
          XM_005261765.1 XP_005261822.1 12 1732
          XM_005261766.1 XP_005261823.1 12 1827
          XM_006724332.3 XP_006724395.1 12 1736
          XM_011530422.1 XP_011528724.1 12 1611
          XM_011530423.1 XP_011528725.1 13 1847
          XM_011530424.1 XP_011528726.1 14 1971
          XM_011530425.1 XP_011528727.1 12 1671
          XM_011530426.2 XP_011528728.1 12 1735
          XM_017028981.1 XP_016884470.1 11 1665
          XM_017028982.1 XP_016884471.1 11 1570
          XM_017028983.1 XP_016884472.1 13 1809
          XM_017028984.1 XP_016884473.1 12 1685
          XM_017028985.1 XP_016884474.1 11 1503
          XM_017028986.1 XP_016884475.1 11 1573
          GenBank mRNA AF064594.1 - 12 1760
          AF102988.1 - 10 1462
          AF102989.1 - 11 1624
          AK001290.1 - 1049
          AK096521.1 - 11 1293
          AK128862.1 - 2 2123
          AK290825.1 - 11 1555
          AK291212.1 - 12 1746
          AK308308.1 - 11 1566
          AL080187.1 - 12 1760
          BC036742.1 - 12 1754 5166749
          BC051904.1 - 11 1534 5175768
          CR456543.1 - 12 1688

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          phospholipase

          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of protein phosphorylation
          chemotaxis
          positive regulation of cytosolic calcium ion concentration
          memory
          urinary bladder smooth muscle contraction
          lipid catabolic process
          antibacterial humoral response
          cardiolipin biosynthetic process
          response to endoplasmic reticulum stress
          positive regulation of insulin secretion involved in cellular response to glucose stimulus
          phosphatidylcholine acyl-chain remodeling
          phosphatidylethanolamine acyl-chain remodeling
          Fc-gamma receptor signaling pathway involved in phagocytosis
          positive regulation of vasodilation
          positive regulation of exocytosis
          negative regulation of synaptic transmission, glutamatergic
          maternal process involved in female pregnancy
          positive regulation of protein kinase C signaling
          positive regulation of release of cytochrome c from mitochondria
          positive regulation of arachidonic acid secretion
          regulation of store-operated calcium channel activity
          positive regulation of ceramide biosynthetic process
          phospholipase A2 activity
          calmodulin binding
          hydrolase activity
          protein kinase binding
          ATP-dependent protein binding
          calcium-independent phospholipase A2 activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-nmghx:80/100.66.79.204:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0