Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other YWHAE SIRNA Assays ›
          Gene Symbol:
          YWHAE YWHAEP1
          Gene Name:
          tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein epsilon
          tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein epsilon pseudogene 1
          Gene Aliases:
          14-3-3E, HEL2, KCIP-1, MDCR, MDS
          -
          Chromosome Location:
          Chr.17: 1344490 - 1400375 on Build GRCh38
          Chromosome Location:
          Chr.7: 64433693 - 64435520 on Build GRCh38
          UniGene ID:
          Hs.513851
          Hs.728871
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          siRNA ID s16
          Validated

          Purification
          Size
          Availability Inventoried
          Catalog # 4390824
          Price 7,898.00
          Your Price
          Online offer:
          7898.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: YWHAE
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_006761.4 NP_006752.1 2 387
          NR_024058.1 3 420
          XM_005256784.3 XP_005256841.1 2 500
          XM_017025005.1 XP_016880494.1 3 370
          GenBank mRNA AK128785.1 - 2 345
          AK295260.1 - 3 255
          AK295997.1 - 2 332
          AK296050.1 - 2 331
          AK316185.1 - 3 378
          AY883089.1 - 3 347
          BC000179.2 - 2 267 2900956
          BC001440.2 - 2 313 3139004
          BM755263.1 - 2 350
          BT007161.1 - 2 235
          DC365395.1 - 2 388
          EU794581.1 - 2 313
          U20972.1 - 2 319
          U28936.1 - 2 235
          U43399.1 - 2 294
          U43430.1 - 2 294
          U54778.1 - 2 295
          Gene Symbol: YWHAEP1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_029404.1 1 407
          GenBank mRNA DB053774.1 - 1 366

          Back To Top

          More Information


          Matching Taqman Gene Expression Assay:


          Hs00356749_g1

          Panther Classification:

          Molecular Function -

          scaffold/adaptor protein

          Gene Ontology Categories:

          Function(s) Process(es)

          G2/M transition of mitotic cell cycle
          neuron migration
          regulation of heart rate by hormone
          protein targeting
          viral process
          substantia nigra development
          hippocampus development
          cerebral cortex development
          hippo signaling
          intracellular signal transduction
          negative regulation of cysteine-type endopeptidase activity involved in apoptotic process
          regulation of membrane repolarization
          membrane organization
          membrane repolarization during cardiac muscle cell action potential
          regulation of heart rate by cardiac conduction
          cell-cell adhesion
          regulation of cellular response to heat
          positive regulation of protein insertion into mitochondrial membrane involved in apoptotic signaling pathway
          regulation of potassium ion transmembrane transporter activity
          negative regulation of peptidyl-serine dephosphorylation
          protein binding
          potassium channel regulator activity
          enzyme binding
          protein domain specific binding
          MHC class II protein complex binding
          ubiquitin protein ligase binding
          histone deacetylase binding
          ion channel binding
          poly(A) RNA binding
          protein heterodimerization activity
          phosphoserine binding
          phosphoprotein binding
          cadherin binding involved in cell-cell adhesion

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0