Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Rictor | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008760806.1 | XP_008759028.1 | 2 | 1459 | ||
XM_008760807.1 | XP_008759029.1 | 5 | 369 | |||
XM_008760808.1 | XP_008759030.1 | 4 | 332 |