Search
Search

| Gene Symbol: Ttc30a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001029918.1 | NP_001025089.1 | 1 | 1521 | ||
| GenBank mRNA | BC098051.1 | - | 1 | 1521 | 7457462 | |
| Gene Symbol: Ttc30a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271494.1 | NP_001258423.1 | 1718 | |||