Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PPFIBP2 SIRNA Assays ›
Gene Symbol:
PPFIBP2
Gene Name:
PPFIA binding protein 2
Gene Aliases:
Cclp1
Chromosome Location:
Chr.11: 7513765 - 7667329 on Build GRCh38
UniGene ID:
Hs.655714
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s16131
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
5,366.00
Your Price
Online offer:
5366.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PPFIBP2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001256568.1
NP_001243497.1
13
1293
NM_001256569.1
NP_001243498.1
12
1273
NM_003621.3
NP_003612.2
16
1870
XM_005253177.2
XP_005253234.1
16
1865
XM_006718349.2
XP_006718412.1
17
1898
XM_006718350.2
XP_006718413.1
17
1898
XM_006718351.2
XP_006718414.1
15
1706
XM_006718352.2
XP_006718415.1
14
1673
XM_006718353.2
XP_006718416.1
13
1265
XM_011520413.2
XP_011518715.1
18
1886
XM_011520414.2
XP_011518716.1
17
1613
XM_011520415.2
XP_011518717.1
17
1898
XM_011520416.2
XP_011518718.1
17
1898
XM_011520417.2
XP_011518719.1
15
1753
XM_011520418.2
XP_011518720.1
14
1350
XM_011520419.2
XP_011518721.1
15
1406
XM_011520420.2
XP_011518722.1
15
1349
XM_017018443.1
XP_016873932.1
17
1853
XM_017018444.1
XP_016873933.1
15
1706
XM_017018445.1
XP_016873934.1
14
1720
XM_017018446.1
XP_016873935.1
14
1673
XM_017018447.1
XP_016873936.1
13
2966
GenBank mRNA
AF034803.1
-
13
1198
AK293669.1
-
14
1318
AK294554.1
-
5
412
AK296718.1
-
13
1293
AK301978.1
-
14
1290
AK308398.1
-
16
1636
AK311048.1
-
6
439
AL834426.1
-
15
1499
BC021714.1
-
16
1818
4826610
CR749258.1
-
14
1308
Back To Top
More Information
Gene Ontology Categories:
Process(es)
molecular_function
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-55658555d6-qfdf2:80/100.66.76.55:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline