Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Sox17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107902.1 | NP_001101372.1 | 4 | 1887 | ||
XM_006237809.2 | XP_006237871.1 | 4 | 2887 | |||
XM_006237812.2 | XP_006237874.1 | 4 | 2706 | |||
XM_008763533.1 | XP_008761755.1 | 5 | 2556 | |||
XM_008763534.1 | XP_008761756.1 | 5 | 2559 | |||
XM_008763535.1 | XP_008761757.1 | 5 | 2306 | |||
XM_008763536.1 | XP_008761758.1 | 5 | 2303 | |||
XM_008763537.1 | XP_008761759.1 | 4 | 2015 | |||
XM_008763538.1 | XP_008761760.1 | 3 | 2041 |