Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Prpf39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003750171.2 | XP_003750219.1 | 6 | 1034 | ||
XM_006240140.2 | XP_006240202.1 | 6 | 1542 | |||
XM_006240141.2 | XP_006240203.1 | 7 | 1439 | |||
XM_006240142.2 | XP_006240204.1 | 6 | 1104 | |||
XM_006240143.2 | XP_006240205.1 | 6 | 752 | |||
XM_006240144.2 | XP_006240206.1 | 5 | 688 | |||
GenBank mRNA | BC166472.1 | - | 5 | 1664 |