Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MKNK1 SIRNA Assays ›
Gene Symbol:
MKNK1
Gene Name:
MAP kinase interacting serine/threonine kinase 1
Gene Aliases:
MNK1
Chromosome Location:
Chr.1: 46557407 - 46607140 on Build GRCh38
UniGene ID:
Hs.371594
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s16320
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
5,366.00
Your Price
Online offer:
5366.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MKNK1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001135553.2
NP_001129025.1
4, 5
479
NM_003684.5
NP_003675.2
4, 5
479
NM_198973.3
NP_945324.1
4, 5
437
NR_024174.2
4, 5
479
NR_024176.2
5, 6
580
XM_006711000.1
XP_006711063.1
4, 5
432
XM_006711001.2
XP_006711064.1
5, 6
682
XM_006711002.2
XP_006711065.1
4, 5
5907
XM_006711003.1
XP_006711066.1
5, 6
493
XM_006711013.1
XP_006711076.1
4, 5
431
XM_006711014.1
XP_006711077.1
4, 5
422
XM_006711015.1
XP_006711078.1
5, 6
493
XM_017002655.1
XP_016858144.1
5, 6
6253
XM_017002656.1
XP_016858145.1
5, 6
6052
XM_017002659.1
XP_016858148.1
4, 5
451
XM_017002660.1
XP_016858149.1
4, 5
5907
XM_017002661.1
XP_016858150.1
5, 6
672
XM_017002662.1
XP_016858151.1
5, 6
493
XM_017002663.1
XP_016858152.1
5, 6
6253
XM_017002664.1
XP_016858153.1
4, 5
5907
XM_017002666.1
XP_016858155.1
5, 6
3382
XM_017002667.1
XP_016858156.1
3, 4
958
GenBank mRNA
AB000409.1
-
4, 5
422
AB451300.1
-
3, 4
235
AB451432.1
-
3, 4
235
AK096423.1
-
5, 6
524
AK290456.1
-
4, 5
399
AK298839.1
-
4, 5
453
AK304005.1
-
4, 5
502
AY355461.1
-
3, 4
235
BC002755.2
-
4, 5
399
3629765
BG722049.1
-
4, 5
442
BX538193.1
-
3, 4
303
BX648559.1
-
5, 6
488
DA435648.1
-
4, 5
424
DA502363.1
-
4, 5
452
DC366212.1
-
4, 5
453
DC376088.1
-
4, 5
409
Back To Top
More Information
Panther Classification:
Molecular Function -
non-receptor serine/threonine protein kinase
Gene Ontology Categories:
Function(s)
Process(es)
regulation of translational initiation
protein phosphorylation
peptidyl-serine phosphorylation
intracellular signal transduction
protein autophosphorylation
protein serine/threonine kinase activity
calmodulin-dependent protein kinase activity
protein binding
calmodulin binding
ATP binding
calcium-dependent protein serine/threonine kinase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0