Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ?
立即註冊
產品
Antibodies
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Cell Culture Media
Chemicals
Western Blot Products
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Greener Products
查看所有產品類別
應用
生物製程
細胞培養和轉染
食品和飲料
實驗室解決方案
製藥和生物製藥
即時定量PCR
半導體分析
臨床和診斷
數位解決方案
分子光譜、同位素與元素分析
查看所有應用與技術
服務
360° CDMO 和 CRO 服務
CDMO 服务
临床试验 CRO 服务
客製化服務
企業服務
金融與租賃服務
儀器服務
實驗室資訊學
授權與商業供應
培訓服務
查看所有服務
支援
線上訂購協助
數位解決方案
產品支持
技術相關資訊
培訓與教育
查看所有協助與支援
熱門商品
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
特價商品
聯絡我們
快速訂購
訂單狀態與貨物追蹤
文件和證書
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
訂單查詢
快速訂購
登入
登入
還沒有帳戶嗎 ?
立即註冊
帳戶
訂單
Connect: Lab, Data, Apps
客製化產品與項目
儀器管理
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MKNK1 SIRNA Assays ›
Gene Symbol:
MKNK1
Gene Name:
MAP kinase interacting serine/threonine kinase 1
Gene Aliases:
MNK1
Chromosome Location:
Chr.1: 46557407 - 46607140 on Build GRCh38
UniGene ID:
Hs.371594
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s16321
Validated
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
1 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
1 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
1 umol
10 umol
Availability
Inventoried
Catalog #
4390824
Price
7,898.00
Your Price
Online offer:
7898.0
Check your price ›
新增到購物車
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MKNK1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001135553.2
NP_001129025.1
5
515
NM_003684.5
NP_003675.2
5
515
NM_198973.3
NP_945324.1
5
473
NR_024174.2
5
515
NR_024176.2
6
616
XM_006711000.1
XP_006711063.1
5
468
XM_006711001.2
XP_006711064.1
6
718
XM_006711002.2
XP_006711065.1
5
5943
XM_006711003.1
XP_006711066.1
6
529
XM_006711006.3
XP_006711069.1
4
3258
XM_006711007.1
XP_006711070.1
4
360
XM_006711009.3
XP_006711072.1
4
5845
XM_006711013.1
XP_006711076.1
5
467
XM_006711014.1
XP_006711077.1
5
458
XM_006711015.1
XP_006711078.1
6
529
XM_017002652.1
XP_016858141.1
5
6191
XM_017002653.1
XP_016858142.1
6
816
XM_017002654.1
XP_016858143.1
6
3668
XM_017002655.1
XP_016858144.1
6
6289
XM_017002656.1
XP_016858145.1
6
6088
XM_017002657.1
XP_016858146.1
5
5990
XM_017002658.1
XP_016858147.1
5
6191
XM_017002659.1
XP_016858148.1
5
487
XM_017002660.1
XP_016858149.1
5
5943
XM_017002661.1
XP_016858150.1
6
708
XM_017002662.1
XP_016858151.1
6
529
XM_017002663.1
XP_016858152.1
6
6289
XM_017002664.1
XP_016858153.1
5
5943
XM_017002665.1
XP_016858154.1
4
360
XM_017002666.1
XP_016858155.1
6
3418
XM_017002667.1
XP_016858156.1
4
994
XM_017002668.1
XP_016858157.1
5
6191
GenBank mRNA
AB000409.1
-
5
458
AB451300.1
-
4
271
AB451432.1
-
4
271
AK096423.1
-
6
560
AK290456.1
-
5
435
AK298839.1
-
5
489
AK304005.1
-
5
538
AK310566.1
-
4
360
AY355461.1
-
4
271
BC002755.2
-
5
435
3629765
BG722049.1
-
5
478
BX538193.1
-
4
339
BX648559.1
-
6
524
DA435648.1
-
5
460
DA502363.1
-
5
488
DC366212.1
-
5
489
DC376088.1
-
5
445
Back To Top
More Information
Matching Taqman Gene Expression Assay:
Hs00374376_m1
Panther Classification:
Molecular Function -
non-receptor serine/threonine protein kinase
Gene Ontology Categories:
Function(s)
Process(es)
regulation of translational initiation
protein phosphorylation
peptidyl-serine phosphorylation
intracellular signal transduction
protein autophosphorylation
protein serine/threonine kinase activity
calmodulin-dependent protein kinase activity
protein binding
calmodulin binding
ATP binding
calcium-dependent protein serine/threonine kinase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0