Search Thermo Fisher Scientific
Gene Symbol: Smug1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177934.3 | NP_808795.1 | 2 | 263 | ||
XM_006242399.2 | XP_006242461.1 | 1 | 84 | |||
XM_008765749.1 | XP_008763971.1 | 2 | 140 | |||
GenBank mRNA | AY191521.1 | - | 2 | 268 |