Search
Search

Gtsf1| Gene Symbol: Gtsf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001079707.1 | NP_001073175.1 | 6 | 552 | ||
| XM_006242403.2 | XP_006242465.1 | 7 | 524 | |||
| XM_008765750.1 | XP_008763972.1 | 7 | 514 | |||
| XM_008765751.1 | XP_008763973.1 | 7 | 516 | |||
| XM_008765752.1 | XP_008763974.1 | 7 | 585 | |||
| GenBank mRNA | BC128778.1 | - | 6 | 552 | 7134533 | |