Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pigu | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181637.1 | NP_853668.1 | 5 | 385 | ||
XM_006235325.2 | XP_006235387.1 | 6 | 636 | |||
XM_008762347.1 | XP_008760569.1 | 3 | 733 | |||
GenBank mRNA | AB086841.1 | - | 5 | 420 | ||
BC097269.1 | - | 6 | 551 |