Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Alg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108264.1 | NP_001101734.1 | 4 | 449 | ||
XM_006245802.2 | XP_006245864.1 | 4 | 485 | |||
XM_008767491.1 | XP_008765713.1 | 4 | 291 | |||
XM_008767492.1 | XP_008765714.1 | 3 | 1349 |