Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Pelp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024270.2 | NP_001019441.2 | 12 | 1296 | ||
GenBank mRNA | AY970831.2 | - | 12 | 1294 | ||
BC101890.1 | - | 12 | 1296 | 7930679 |