Search
Search

Nt5c3b| Gene Symbol: Nt5c3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007723.2 | NP_001007724.2 | 7 | 576 | ||
| XM_008768083.1 | XP_008766305.1 | 5 | 394 | |||
| XM_008768084.1 | XP_008766306.1 | 5 | 394 | |||
| GenBank mRNA | BC078963.1 | - | 7 | 576 | 7111222 | |