Search
Search

Lnx1| Gene Symbol: Lnx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108358.1 | NP_001101828.1 | 5 | 1031 | ||
| XM_006250902.2 | XP_006250964.1 | 5 | 1025 | |||
| XM_006250903.2 | XP_006250965.1 | 4 | 3063 | |||
| XM_006250904.2 | XP_006250966.1 | 4 | 3033 | |||
| GenBank mRNA | EU219404.1 | - | 4 | 874 | ||
| EU219405.1 | - | 4 | 874 | |||
| EU219406.1 | - | 4 | 742 | |||