Search
Search

Entpd4| Gene Symbol: Entpd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108384.1 | NP_001101854.1 | 5 | 822 | ||
| XM_006252275.2 | XP_006252337.1 | 10 | 1575 | |||
| XM_006252276.2 | XP_006252338.1 | 10 | 1978 | |||
| XM_006252277.2 | XP_006252339.1 | 10 | 1551 | |||