Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PER3 SIRNA Assays ›
Gene Symbol:
PER3
Gene Name:
period circadian clock 3
Gene Aliases:
FASPS3, GIG13
Chromosome Location:
Chr.1: 7784285 - 7845181 on Build GRCh38
UniGene ID:
Hs.162200
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s16928
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
392.00
Your Price
Online offer:
380.65
380.65
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PER3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001289861.1
NP_001276790.1
18
2828
NM_001289862.1
NP_001276791.1
18
2828
NM_001289863.1
NP_001276792.1
18
2807
NM_001289864.1
NP_001276793.1
18
2889
NM_016831.2
NP_058515.1
18
2804
XM_005263524.3
XP_005263581.1
17
2703
XM_011542385.2
XP_011540687.1
17
2703
XM_011542390.2
XP_011540692.1
17
2682
XM_017002723.1
XP_016858212.1
17
2700
XM_017002724.1
XP_016858213.1
17
2700
XM_017002725.1
XP_016858214.1
17
2682
XM_017002726.1
XP_016858215.1
17
2679
XM_017002727.1
XP_016858216.1
19
3022
XM_017002728.1
XP_016858217.1
17
2764
XM_017002729.1
XP_016858218.1
15
2204
XM_017002730.1
XP_016858219.1
17
2764
XM_017002731.1
XP_016858220.1
17
2743
XM_017002732.1
XP_016858221.1
16
2792
XM_017002733.1
XP_016858222.1
19
2929
XM_017002734.1
XP_016858223.1
20
3071
XM_017002735.1
XP_016858224.1
19
2929
XM_017002736.1
XP_016858225.1
20
3050
XM_017002737.1
XP_016858226.1
14
2187
XM_017002738.1
XP_016858227.1
14
2193
GenBank mRNA
AB047686.1
-
17
2788
AK307760.1
-
17
2682
AK308303.1
-
18
2877
AL157954.1
-
17
2715
AY493418.1
-
17
2611
BC111010.1
-
1
72
BC144137.1
-
17
2793
BC146781.1
-
17
2788
9007235
Back To Top
More Information
Panther Classification:
Molecular Function -
transcription cofactor
gene-specific transcriptional regulator
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
transcription, DNA-templated
circadian regulation of gene expression
regulation of circadian sleep/wake cycle, sleep
protein stabilization
protein binding
kinase binding
ubiquitin protein ligase binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-2rf75:80/100.66.79.83:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0