Search Thermo Fisher Scientific
- ์ฃผ๋ฌธ ํํฉ
- ๋น ๋ฅธ์ฃผ๋ฌธ
-
ํ์์ด ์๋์ ๊ฐ์? ๊ณ์ ์์ฑํ๊ธฐ
Search Thermo Fisher Scientific
Gene Symbol: Itgb3bp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013213.1 | NP_001013231.1 | 6, 7 | 755 | ||
XM_008763882.1 | XP_008762104.1 | 4, 5 | 572 | |||
XM_008763883.1 | XP_008762105.1 | 7, 8 | 497 | |||
XM_008763884.1 | XP_008762106.1 | 6, 7 | 451 | |||
GenBank mRNA | BC086363.1 | - | 6, 7 | 755 | 7304501 |