Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ARHGEF7 SIRNA Assays ›
          Gene Symbol:
          ARHGEF7
          Gene Name:
          Rho guanine nucleotide exchange factor 7
          Gene Aliases:
          BETA-PIX, COOL-1, COOL1, Nbla10314, P50, P50BP, P85, P85COOL1, P85SPR, PAK3, PIXB
          Chromosome Location:
          Chr.13: 111114619 - 111305734 on Build GRCh38
          UniGene ID:
          Hs.508738
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s16950

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 392.00
          Your Price
          Online offer: 380.65
          380.65
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ARHGEF7
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001113511.2 NP_001106983.1 16 2580
          NM_001113512.2 NP_001106984.1 14 2430
          NM_001113513.1 NP_001106985.1 14 1522
          NM_001320851.1 NP_001307780.1 13 1303
          NM_001320852.1 NP_001307781.1 15 2517
          NM_001320853.1 NP_001307782.1 14 1428
          NM_001320854.1 NP_001307783.1 10 1042
          NM_003899.3 NP_003890.1 14 1635
          NM_145735.3 NP_663788.1 15 2517
          XM_006719956.3 XP_006720019.1 15 1645
          XM_011521133.2 XP_011519435.1 14 1566
          XM_017020814.1 XP_016876303.1 15 1645
          XM_017020815.1 XP_016876304.1 13 1638
          XM_017020816.1 XP_016876305.1 14 1413
          XM_017020817.1 XP_016876306.1 13 1638
          XM_017020818.1 XP_016876307.1 15 1513
          XM_017020819.1 XP_016876308.1 14 1522
          XM_017020820.1 XP_016876309.1 14 1645
          XM_017020821.1 XP_016876310.1 14 1340
          XM_017020822.1 XP_016876311.1 13 1336
          XM_017020823.1 XP_016876312.1 13 2113
          XM_017020824.1 XP_016876313.1 15 1513
          XM_017020825.1 XP_016876314.1 13 1264
          XM_017020826.1 XP_016876315.1 15 1513
          XM_017020827.1 XP_016876316.1 13 1337
          XM_017020828.1 XP_016876317.1 11 961
          XM_017020829.1 XP_016876318.1 11 961
          GenBank mRNA AB075521.1 - 5 376
          AB177848.1 - 15 2518
          AB177849.1 - 12 1241
          AK295472.1 - 10 967
          AK300157.1 - 12 1225
          AK300272.1 - 14 1415
          AL834228.1 - 15 1701
          BC033905.1 - 14 1627 5262912
          BC050521.1 - 14 1494
          BC060776.1 - 16 1922 5271977
          D63476.1 - 14 1611

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          guanyl-nucleotide exchange factor

          Gene Ontology Categories:

          Function(s) Process(es)

          hematopoietic progenitor cell differentiation
          Golgi organization
          signal transduction
          nervous system development
          positive regulation of fibroblast migration
          lamellipodium assembly
          positive regulation of protein binding
          regulation of Rho protein signal transduction
          intracellular signal transduction
          negative regulation of epidermal growth factor receptor signaling pathway
          positive regulation of apoptotic process
          positive regulation of GTPase activity
          ephrin receptor signaling pathway
          focal adhesion assembly
          regulation of small GTPase mediated signal transduction
          positive regulation of growth hormone secretion
          positive regulation of substrate adhesion-dependent cell spreading
          regulation of GTP binding
          positive regulation of lamellipodium morphogenesis
          guanyl-nucleotide exchange factor activity
          Rho guanyl-nucleotide exchange factor activity
          protein binding
          protein kinase binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-2rf75:80/100.66.79.83:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0