Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other AP1G2 SIRNA Assays ›
Gene Symbol:
AP1G2
Gene Name:
adaptor related protein complex 1 gamma 2 subunit
Gene Aliases:
G2AD
Chromosome Location:
Chr.14: 23559563 - 23569056 on Build GRCh38
UniGene ID:
Hs.343244
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s17031
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
5,366.00
Your Price
Online offer:
5366.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: AP1G2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001282474.1
NP_001269403.1
7, 8
1078
NM_001282475.1
NP_001269404.1
7, 8
928
NM_003917.4
NP_003908.1
9, 10
1032
XM_005268167.3
XP_005268224.1
8, 9
1155
XM_005268168.4
XP_005268225.1
8, 9
1197
XM_005268169.3
XP_005268226.1
8, 9
1154
XM_005268170.3
XP_005268227.1
8, 9
1154
XM_005268172.3
XP_005268229.1
9, 10
983
XM_005268173.3
XP_005268230.1
9, 10
983
XM_005268174.3
XP_005268231.1
7, 8
1030
XM_005268175.3
XP_005268232.1
7, 8
1044
XM_005268177.3
XP_005268234.1
8, 9
1010
XM_005268178.3
XP_005268235.1
8, 9
901
XM_005268179.3
XP_005268236.1
8, 9
1006
XM_005268180.4
XP_005268237.1
7, 8
1248
XM_005268182.3
XP_005268239.1
8, 9
877
XM_006720301.3
XP_006720364.1
8, 9
1252
XM_011537283.2
XP_011535585.1
7, 8
1101
XM_011537286.2
XP_011535588.1
1, 2
25
XM_017021740.1
XP_016877229.1
7, 8
1101
XM_017021741.1
XP_016877230.1
5, 6
1005
XM_017021742.1
XP_016877231.1
7, 8
1072
XM_017021743.1
XP_016877232.1
7, 8
1101
XM_017021744.1
XP_016877233.1
9, 10
984
XM_017021745.1
XP_016877234.1
9, 10
984
XM_017021746.1
XP_016877235.1
8, 9
877
XM_017021747.1
XP_016877236.1
8, 9
2269
XM_017021748.1
XP_016877237.1
7, 8
770
GenBank mRNA
AB015318.1
-
9, 10
974
AF068706.1
-
8, 9
1692
AK123354.1
-
1, 2
79
AK304682.1
-
7, 8
763
BC051833.1
-
6, 7
869
5589077
BX161425.1
-
7, 8
1011
BX247963.1
-
6, 7
737
Back To Top
More Information
Panther Classification:
Molecular Function -
membrane traffic protein
Gene Ontology Categories:
Function(s)
Process(es)
intracellular protein transport
viral process
vesicle-mediated transport
protein binding
protein transporter activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-prz7w:80/100.66.75.107:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0