Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other BCL10 SIRNA Assays ›
          Gene Symbol:
          BCL10
          Gene Name:
          B-cell CLL/lymphoma 10
          Gene Aliases:
          CARMEN, CIPER, CLAP, IMD37, c-E10, mE10
          Chromosome Location:
          Chr.1: 85265776 - 85276904 on Build GRCh38
          UniGene ID:
          Hs.193516
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s17057

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 5,366.00
          Your Price
          Online offer:
          5366.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: BCL10
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001320715.1 NP_001307644.1 2 800
          NM_003921.4 NP_003912.1 2 800
          XM_011542397.2 XP_011540699.1 2 669
          XM_011542398.2 XP_011540700.1 2 674
          XM_011542399.2 XP_011540701.1 2 822
          GenBank mRNA AF057700.1 - 2 494
          AF082283.1 - 3 954
          AF100338.1 - 2 495
          AF105066.1 - 2 248
          AF127386.1 - 2 248
          AF134395.1 - 2 493
          AJ006288.1 - 3 938
          AK291346.1 - 2 458
          BC053617.1 - 2 370 6143423
          EF189176.1 - 2 353

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          B cell apoptotic process
          neural tube closure
          stimulatory C-type lectin receptor signaling pathway
          toll-like receptor signaling pathway
          response to molecule of bacterial origin
          adaptive immune response
          negative regulation of mature B cell apoptotic process
          cellular defense response
          I-kappaB kinase/NF-kappaB signaling
          cell death
          response to fungus
          immunoglobulin mediated immune response
          positive regulation of protein ubiquitination
          response to food
          positive regulation of mast cell cytokine production
          Fc-epsilon receptor signaling pathway
          lymphotoxin A biosynthetic process
          interleukin-6 biosynthetic process
          positive regulation of phosphorylation
          positive regulation of I-kappaB kinase/NF-kappaB signaling
          positive regulation of cysteine-type endopeptidase activity involved in apoptotic process
          innate immune response
          positive regulation of interleukin-8 biosynthetic process
          positive regulation of transcription, DNA-templated
          T cell receptor signaling pathway
          regulation of T cell receptor signaling pathway
          positive regulation of T cell activation
          positive regulation of NF-kappaB transcription factor activity
          protein oligomerization
          protein homooligomerization
          protein heterooligomerization
          T cell apoptotic process
          cellular response to mechanical stimulus
          positive regulation of extrinsic apoptotic signaling pathway
          protease binding
          transcription coactivator activity
          protein binding
          protein C-terminus binding
          transcription factor binding
          kinase activator activity
          enzyme binding
          kinase binding
          protein kinase binding
          ubiquitin protein ligase binding
          identical protein binding
          ubiquitin binding
          protein kinase B binding
          protein self-association
          NF-kappaB binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-mkfsp:80/100.66.78.65:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0