Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ATP1A1 SIRNA Assays ›
          Gene Symbol:
          ATP1A1
          Gene Name:
          ATPase Na+/K+ transporting subunit alpha 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 116373173 - 116404774 on Build GRCh38
          UniGene ID:
          Hs.371889
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s1718
          Validated

          Purification
          Size
          Availability Inventoried
          Catalog # 4390824
          Price 7,898.00
          Your Price
          Online offer:
          7898.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ATP1A1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000701.7 NP_000692.2 7 995
          NM_001160233.1 NP_001153705.1 7 1019
          NM_001160234.1 NP_001153706.1 7 828
          XM_017001360.1 XP_016856849.1 8 4214
          XM_017001361.1 XP_016856850.1 8 3556
          GenBank mRNA AK295095.1 - 7 1018
          AK296362.1 - 7 828
          AK296384.1 - 7 954
          AK308418.1 - 7 760
          AK314777.1 - 7 840
          BC003077.2 - 7 908 3507685
          BC050359.1 - 7 906 6065209
          J03007.1 - 2 65
          U16798.1 - 6 656

          Back To Top

          More Information


          Matching Taqman Gene Expression Assay:


          Hs00167556_m1

          Panther Classification:

          Molecular Function -

          primary active transporter

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of the force of heart contraction
          regulation of sodium ion transport
          cellular sodium ion homeostasis
          regulation of blood pressure
          potassium ion import
          establishment or maintenance of transmembrane electrochemical gradient
          ATP hydrolysis coupled proton transport
          dephosphorylation
          cellular potassium ion homeostasis
          negative regulation of glucocorticoid biosynthetic process
          ion transmembrane transport
          sodium ion export from cell
          response to drug
          negative regulation of heart contraction
          positive regulation of heart contraction
          positive regulation of striated muscle contraction
          relaxation of cardiac muscle
          cardiac muscle contraction
          membrane hyperpolarization
          cellular response to mechanical stimulus
          cellular response to steroid hormone stimulus
          regulation of cardiac muscle cell contraction
          membrane repolarization
          membrane repolarization during cardiac muscle cell action potential
          cell communication by electrical coupling involved in cardiac conduction
          response to glycoside
          regulation of cardiac conduction
          potassium ion import across plasma membrane
          sodium:potassium-exchanging ATPase activity
          protein binding
          ATP binding
          phosphatase activity
          protein kinase binding
          protein domain specific binding
          ankyrin binding
          potassium ion binding
          sodium ion binding
          ADP binding
          phosphatidylinositol 3-kinase binding
          chaperone binding
          steroid hormone binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0