Search
Search

CCRL2| Gene Symbol: CCRL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001130910.1 | NP_001124382.1 | 2 | 336 | ||
| NM_003965.4 | NP_003956.2 | 2 | 472 | |||
| XM_011534208.1 | XP_011532510.1 | 3 | 416 | |||
| XM_011534209.1 | XP_011532511.1 | 3 | 339 | |||
| XM_017007436.1 | XP_016862925.1 | 2 | 332 | |||
| GenBank mRNA | AF015524.1 | - | 2 | 270 | ||
| AF015525.1 | - | 2 | 514 | |||
| AJ344142.1 | - | 1 | 187 | |||
| AK296673.1 | - | 2 | 336 | |||
| AK313312.1 | - | 2 | 260 | |||
| BC071682.1 | - | 2 | 425 | 5182006 | ||
| BC096075.1 | - | 2 | 252 | 40000578 | ||
| BC096076.3 | - | 2 | 252 | 40000580 | ||