Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other P2RX6 SIRNA Assays ›
Gene Symbol:
P2RX6
Gene Name:
purinergic receptor P2X 6
Gene Aliases:
P2RXL1, P2X6, P2XM
Chromosome Location:
Chr.22: 21009699 - 21028013 on Build GRCh38
UniGene ID:
Hs.113275
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s17429
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
5,366.00
Your Price
Online offer:
5366.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: P2RX6
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001159554.1
NP_001153026.1
2
174
NM_005446.3
NP_005437.2
2
230
XM_005261819.3
XP_005261876.1
2
954
XM_011530498.2
XP_011528800.1
2
955
XM_011530499.2
XP_011528801.1
2
956
XM_011530500.2
XP_011528802.1
2
877
XM_011530501.2
XP_011528803.1
2
230
XM_011530502.2
XP_011528804.1
2
955
XM_017029066.1
XP_016884555.1
2
955
XM_017029067.1
XP_016884556.1
2
954
XM_017029068.1
XP_016884557.1
2
954
XM_017029069.1
XP_016884558.1
2
875
XM_017029070.1
XP_016884559.1
2
232
XM_017029071.1
XP_016884560.1
2
954
XM_017029072.1
XP_016884561.1
2
187
XM_017029073.1
XP_016884562.1
2
230
XM_017029074.1
XP_016884563.1
2
151
XM_017029075.1
XP_016884564.1
2
244
XM_017029076.1
XP_016884565.1
2
874
GenBank mRNA
AB002058.1
-
2
245
AF065385.1
-
2
244
AK309079.1
-
2
230
BC033488.1
-
2
214
5176046
BC047287.1
-
2
213
5198937
BC063553.1
-
2
215
BC064805.1
-
2
174
BC109209.2
-
2
157
40006731
BI753477.1
-
2
174
CR456535.1
-
2
236
DA333742.1
-
2
230
Back To Top
More Information
Panther Classification:
Molecular Function -
ligand-gated ion channel
Gene Ontology Categories:
Function(s)
Process(es)
transport
muscle contraction
signal transduction
blood coagulation
response to ATP
purinergic nucleotide receptor signaling pathway
protein homooligomerization
protein heterooligomerization
cation transmembrane transport
purinergic nucleotide receptor activity
transmembrane signaling receptor activity
extracellular ATP-gated cation channel activity
ATP binding
channel activity
identical protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-55658555d6-7l2qm:80/100.66.75.127:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline