Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Fut10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012360.1 | NP_001012360.1 | 3 | 579 | ||
XM_006253236.2 | XP_006253298.1 | 4 | 964 | |||
GenBank mRNA | AJ879584.1 | - | 3 | 579 | ||
AJ880010.1 | - | 3 | 579 |