Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other AURKB SIRNA Assays ›
Gene Symbol:
AURKB
Gene Name:
aurora kinase B
Gene Aliases:
AIK2, AIM-1, AIM1, ARK2, AurB, IPL1, PPP1R48, STK12, STK5, aurkb-sv1, aurkb-sv2
Chromosome Location:
Chr.17: 8204731 - 8210768 on Build GRCh38
UniGene ID:
Hs.442658
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s17612
Validated
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
1 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
1 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
1 umol
10 umol
Availability
Inventoried
Catalog #
4390824
Price
7,898.00
Your Price
Online offer:
7898.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: AURKB
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001256834.2
NP_001243763.1
7, 8
911
NM_001284526.1
NP_001271455.1
8, 9
987
NM_001313950.1
NP_001300879.1
8, 9
960
NM_001313951.1
NP_001300880.1
8, 9
989
NM_001313952.1
NP_001300881.1
7, 8
975
NM_001313953.1
NP_001300882.1
7, 8
767
NM_001313954.1
NP_001300883.1
8, 9
994
NM_001313955.1
NP_001300884.1
6, 7
630
NM_004217.3
NP_004208.2
8, 9
984
NR_132730.1
8, 9
842
NR_132731.1
6, 7
727
XM_011524070.2
XP_011522372.1
8, 9
837
XM_011524072.2
XP_011522374.1
7, 8
1100
XM_017025307.1
XP_016880796.1
7, 8
1027
XM_017025308.1
XP_016880797.1
7, 8
957
XM_017025309.1
XP_016880798.1
7, 8
865
XM_017025310.1
XP_016880799.1
8, 9
847
XM_017025311.1
XP_016880800.1
4, 5
524
GenBank mRNA
AB011446.1
-
8, 9
925
AB011450.1
-
8, 9
919
AB519677.1
-
7, 8
862
AB519678.1
-
7, 8
766
AB519679.1
-
6, 7
726
AF004022.1
-
8, 9
929
AF008552.1
-
7, 8
862
AF015254.1
-
8, 9
929
AK297976.1
-
7, 8
866
AK314889.1
-
8, 9
933
AY677083.1
-
6, 7
727
BC000442.2
-
8, 9
923
2820719
BC009751.2
-
8, 9
903
3677349
BC013300.2
-
8, 9
943
BC080581.1
-
8, 9
915
2819846
BT019534.1
-
7, 8
862
Back To Top
More Information
Matching Taqman Gene Expression Assay:
Hs00177782_m1
Panther Classification:
Molecular Function -
non-receptor serine/threonine protein kinase
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
negative regulation of B cell apoptotic process
protein phosphorylation
spindle organization
sister chromatid cohesion
aging
cell proliferation
attachment of spindle microtubules to kinetochore
abscission
histone modification
protein sumoylation
anaphase-promoting complex-dependent catabolic process
spindle checkpoint
negative regulation of protein binding
positive regulation of telomere maintenance via telomerase
negative regulation of cytokinesis
positive regulation of cytokinesis
protein localization to kinetochore
cellular response to UV
cleavage furrow formation
protein ubiquitination involved in ubiquitin-dependent protein catabolic process
histone H3-S28 phosphorylation
protein autophosphorylation
mitotic spindle midzone assembly
positive regulation of telomerase activity
regulation of chromosome segregation
regulation of signal transduction by p53 class mediator
positive regulation of telomere capping
protein serine/threonine kinase activity
protein serine/threonine/tyrosine kinase activity
protein binding
ATP binding
histone serine kinase activity
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0