Search
Search

Tsc22d1| Gene Symbol: Tsc22d1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109912.1 | NP_001103382.1 | 2 | 3203 | ||
| NM_013043.2 | NP_037175.1 | 2 | 331 | |||
| XM_006252339.1 | XP_006252401.1 | 3 | 3075 | |||
| GenBank mRNA | BC059146.1 | - | 2 | 342 | ||
| BC127547.1 | - | 2 | 323 | |||
| CB771530.1 | - | 2 | 331 | |||
| L25785.1 | - | 2 | 324 | |||