Search
Search

RGD1565355
Cd36
LOC103690020| Gene Symbol: RGD1565355 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC091104.1 | - | 2323 | 7383442 | ||
| Gene Symbol: Cd36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031561.2 | NP_113749.2 | 2329 | |||
| GenBank mRNA | BC072543.1 | - | 2329 | 7105943 | ||
| Gene Symbol: LOC103690020 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006235962.2 | XP_006236024.1 | 15 | 2755 | ||
| XM_006235963.2 | XP_006236025.1 | 2328 | ||||
| XM_006235964.2 | XP_006236026.1 | 15 | 2771 | |||