Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other SLC9A3R1 SIRNA Assays ›
          Gene Symbol:
          SLC9A3R1
          Gene Name:
          SLC9A3 regulator 1
          Gene Aliases:
          EBP50, NHERF, NHERF-1, NHERF1, NPHLOP2
          Chromosome Location:
          Chr.17: 74748612 - 74769360 on Build GRCh38
          UniGene ID:
          Hs.724482
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s17920
          Validated

          Purification
          Size
          Availability Inventoried
          Catalog # 4390824
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: SLC9A3R1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_004252.4 NP_004243.1 4 1007
          GenBank mRNA AF015926.1 - 4 984
          AF036241.1 - 4 942
          AK026581.1 - 4 990
          AK128474.1 - 5 956
          AK312498.1 - 4 1023
          BC001443.2 - 4 960 3138881
          BC003361.1 - 4 977 2900516
          BC011777.1 - 4 976 3640162
          BC049220.1 - 4 983
          BC053350.1 - 4 967 5218414
          BX648303.1 - 2 2666

          Back To Top

          More Information


          Matching Taqman Gene Expression Assay:


          Hs00188594_m1

          Gene Ontology Categories:

          Function(s) Process(es)

          renal sodium ion transport
          protein complex assembly
          adenylate cyclase-activating dopamine receptor signaling pathway
          sensory perception of sound
          negative regulation of cell proliferation
          regulation of cell shape
          regulation of cell size
          negative regulation of platelet-derived growth factor receptor signaling pathway
          negative regulation of phosphatidylinositol 3-kinase signaling
          Wnt signaling pathway
          cAMP-mediated signaling
          gland morphogenesis
          microvillus assembly
          actin cytoskeleton organization
          negative regulation of cell migration
          cellular phosphate ion homeostasis
          regulation of sodium:proton antiporter activity
          negative regulation of sodium:proton antiporter activity
          bile acid secretion
          glutathione transport
          negative regulation of catenin import into nucleus
          establishment of nucleus localization
          regulation of excretion
          establishment of epithelial cell apical/basal polarity
          regulation of protein kinase activity
          negative regulation of mitotic cell cycle
          establishment of Golgi localization
          negative regulation of protein kinase B signaling
          auditory receptor cell stereocilium organization
          phospholipase C-activating dopamine receptor signaling pathway
          renal absorption
          negative regulation of ERK1 and ERK2 cascade
          establishment of protein localization to plasma membrane
          renal phosphate ion absorption
          positive regulation of intrinsic apoptotic signaling pathway
          receptor binding
          protein binding
          beta-catenin binding
          chloride channel regulator activity
          phosphatase binding
          PDZ domain binding
          beta-2 adrenergic receptor binding
          protein complex binding
          protein complex scaffold
          protein self-association
          dopamine receptor binding
          growth factor receptor binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-vsgnr:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0