Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: TM9SF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004800.2 | NP_004791.1 | 4 | 675 | ||
GenBank mRNA | AK290514.1 | - | 4 | 630 | ||
AK315210.1 | - | 4 | 630 | |||
BC110656.1 | - | 4 | 632 | 3855486 | ||
BC130293.1 | - | 4 | 509 | 40146324 | ||
U81006.1 | - | 4 | 575 |