Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Zcchc17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109267.1 | NP_001102737.1 | 4 | 324 | ||
XM_006238968.2 | XP_006239030.1 | 4 | 344 | |||
XM_006238969.2 | XP_006239031.1 | 3 | 280 | |||
XM_006238970.2 | XP_006239032.1 | 2 | 159 |