Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dcdc2c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195807.1 | NP_001182736.1 | 1, 2 | 376 | ||
XM_006239955.2 | XP_006240017.1 | 1, 2 | 747 | |||
XM_006239956.2 | XP_006240018.1 | 1, 2 | 746 | |||
XM_006239957.2 | XP_006240019.1 | 1, 2 | 746 | |||
XM_006239958.2 | XP_006240020.1 | 1, 2 | 746 | |||
GenBank mRNA | CK596582.1 | - | 1, 2 | 376 |