Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other LOC500959 SIRNA Assays ›
Gene Symbol:
LOC500959
LOC100911515
Tpi1
Gene Name:
triosephosphate isomerase
triosephosphate isomerase-like
triosephosphate isomerase 1
Gene Aliases:
-
-
Tpi
Chromosome Location:
Chr.8: 26341789 - 26342573 on Build Rnor_6.0
Chromosome Location:
Chr.9: 20213596 - 20217278 on Build Rnor_6.0
Chromosome Location:
Chr.4: 157328375 - 157331905 on Build Rnor_6.0
UniGene ID:
Rn.129979
-
Rn.144555
Species:
Rat
Species Specific ID (Flybase ID):
-
-
-
siRNA ID
s179935
Purification
Standard
HPLC
HPLC-IVR
Size
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
Availability
Made To Order
Catalog #
4390771
Price
5,354.00
Your Price
Online offer:
5354.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: LOC500959
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001033072.1
NP_001028244.1
1
730
GenBank mRNA
AY461585.1
-
1
730
Gene Symbol: LOC100911515
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
XM_003750654.3
XP_003750702.2
7
889
Gene Symbol: Tpi1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_022922.2
NP_075211.2
7
733
GenBank mRNA
BC061781.1
-
7
708
BC126087.1
-
7
733
8368624
FQ214607.1
-
7
819
FQ214670.1
-
7
741
FQ214880.1
-
7
746
FQ215213.1
-
6
648
FQ215529.1
-
7
745
FQ215606.1
-
6
621
FQ215810.1
-
7
746
FQ216226.1
-
6
671
FQ216307.1
-
7
632
FQ216584.1
-
7
745
FQ223237.1
-
7
747
FQ224880.1
-
7
744
FQ224920.1
-
7
748
FQ224970.1
-
7
741
L36250.1
-
7
733
Back To Top
More Information
Panther Classification:
Molecular Function -
isomerase
Gene Ontology Categories:
Function(s)
Process(es)
glucose metabolic process
multicellular organism development
glyceraldehyde-3-phosphate metabolic process
carbohydrate metabolic process
gluconeogenesis
glycolytic process
pentose-phosphate shunt
triose-phosphate isomerase activity
isomerase activity
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System