Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other LOC100912132 SIRNA Assays ›
          Gene Symbol:
          LOC100912132 LOC100362967 RGD1561736 RGD1563861 RGD1564963 Rpl10
          Gene Name:
          60S ribosomal protein L10-like
          ribosomal protein 10-like
          similar to ribosomal protein L10
          similar to ribosomal protein L10
          similar to ribosomal protein L10
          ribosomal protein L10
          Gene Aliases:
          -
          -
          -
          -
          -
          -
          Chromosome Location:
          Chr.15: 101505377 - 101505808 on Build Rnor_6.0
          Chromosome Location:
          Chr.2: 238839534 - 238839941 on Build Rnor_6.0
          Chromosome Location:
          Chr.3: 25148527 - 25149104 on Build Rnor_6.0
          Chromosome Location:
          Chr.18: 14731669 - 14732382 on Build Rnor_6.0
          Chromosome Location:
          Chr.7: 137917283 - 137917822 on Build Rnor_6.0
          Chromosome Location:
          Chr.X: 156438251 - 156440461 on Build Rnor_6.0
          UniGene ID:
          -
          -
          -
          -
          -
          Rn.102157
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          -
          -
          -
          -
          -
          siRNA ID s181440

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price 400.00
          Your Price
          Online offer: 388.65
          388.65
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: LOC100912132
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq XM_006252449.2 XP_006252511.2 1 88
          Gene Symbol: LOC100362967
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq XM_006233349.2 XP_006233411.1 1 96
          Gene Symbol: RGD1561736
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq XM_006234137.2 XP_006234199.1 1 267
          Gene Symbol: RGD1563861
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq XM_574109.5 XP_574109.1 1 455
          Gene Symbol: RGD1564963
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq XM_006242289.2 XP_006242351.1 1 225
          Gene Symbol: Rpl10
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_031100.1 NP_112362.1 6 470
          GenBank mRNA BC058467.1 - 6 459 6918973
          FQ210278.1 - 6 454
          FQ210394.1 - 2 172
          FQ211102.1 - 2 172
          FQ212456.1 - 2 172
          FQ217196.1 - 2 172
          FQ217560.1 - 2 172
          FQ217981.1 - 2 172
          FQ218098.1 - 6 454
          FQ220555.1 - 5 822
          FQ221411.1 - 2 172
          FQ221714.1 - 2 172
          FQ221864.1 - 6 454
          FQ222021.1 - 6 454
          FQ222201.1 - 2 172
          FQ222372.1 - 2 172
          FQ222670.1 - 2 172
          FQ222782.1 - 2 172
          FQ222847.1 - 2 172
          FQ223155.1 - 2 172
          FQ223157.1 - 2 172
          FQ224407.1 - 2 172
          FQ224785.1 - 2 172
          FQ226301.1 - 2 172
          FQ228167.1 - 2 172
          FQ229032.1 - 6 453
          FQ229696.1 - 4 581
          FQ231133.1 - 2 172
          FQ233952.1 - 6 454
          FQ235226.1 - 2 172
          X87106.1 - 6 470

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          ribosomal protein

          Gene Ontology Categories:

          Function(s) Process(es)

          translation
          structural constituent of ribosome
          poly(A) RNA binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-green-55658555d6-7l2qm:80/100.66.75.127:80.
          git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.41.0-Offline