Search
Search

KIF23| Gene Symbol: KIF23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001281301.1 | NP_001268230.1 | 13 | 1539 | ||
| NM_004856.6 | NP_004847.2 | 14 | 1676 | |||
| NM_138555.3 | NP_612565.1 | 14 | 1676 | |||
| XM_005254796.2 | XP_005254853.1 | 15 | 1667 | |||
| XM_005254797.2 | XP_005254854.1 | 15 | 1667 | |||
| XM_005254798.2 | XP_005254855.1 | 15 | 1667 | |||
| XM_005254799.2 | XP_005254856.1 | 15 | 1667 | |||
| XM_011522238.1 | XP_011520540.1 | 14 | 1502 | |||
| XM_011522239.2 | XP_011520541.1 | 14 | 1527 | |||
| XM_011522240.1 | XP_011520542.1 | 6 | 521 | |||
| XM_017022733.1 | XP_016878222.1 | 15 | 1667 | |||
| GenBank mRNA | AK303874.1 | - | 10 | 1070 | ||
| AK308322.1 | - | 13 | 1461 | |||
| AK308361.1 | - | 8 | 885 | |||
| BC017705.2 | - | 13 | 1444 | |||
| BC051826.1 | - | 14 | 1618 | 6500787 | ||
| BX477359.1 | - | 5 | 460 | |||
| X67155.2 | - | 13 | 1516 | |||