Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other KIF1A SIRNA Assays ›
          Gene Symbol:
          KIF1A
          Gene Name:
          kinesin family member 1A
          Gene Aliases:
          ATSV, C2orf20, HSN2C, MRD9, SPG30, UNC104
          Chromosome Location:
          Chr.2: 240713764 - 240821025 on Build GRCh38
          UniGene ID:
          Hs.516802
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s1828

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 5,366.00
          Your Price
          Online offer:
          5366.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: KIF1A
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001244008.1 NP_001230937.1 30, 31 3441
          NM_001320705.1 NP_001307634.1 29, 30 3165
          NM_004321.6 NP_004312.2 27, 28 2951
          XM_005247022.1 XP_005247079.1 30, 31 3352
          XM_005247024.1 XP_005247081.1 29, 30 3325
          XM_005247027.1 XP_005247084.1 28, 29 3049
          XM_005247028.2 XP_005247085.1 28, 29 3052
          XM_006712605.1 XP_006712668.1 29, 30 3325
          XM_011511364.1 XP_011509666.1 30, 31 3364
          XM_011511365.1 XP_011509667.1 29, 30 3076
          XM_011511366.2 XP_011509668.1 20, 21 2424
          XM_011511367.1 XP_011509669.1 20, 21 2354
          XM_017004383.1 XP_016859872.1 30, 31 3427
          XM_017004384.1 XP_016859873.1 30, 31 3439
          XM_017004385.1 XP_016859874.1 30, 31 3427
          XM_017004386.1 XP_016859875.1 30, 31 3427
          XM_017004387.1 XP_016859876.1 29, 30 3151
          XM_017004388.1 XP_016859877.1 29, 30 3151
          XM_017004389.1 XP_016859878.1 29, 30 3151
          XM_017004390.1 XP_016859879.1 28, 29 3049
          XM_017004391.1 XP_016859880.1 29, 30 3206
          GenBank mRNA AB209138.1 - 3, 4 250
          AB210029.1 - 30, 31 3441
          AB290172.1 - 30, 31 3441
          BC111780.1 - 27, 28 2919 40037563
          BC111799.1 - 27, 28 2919 40037561
          KJ534875.1 - 27, 28 2891

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          microtubule binding motor protein

          Gene Ontology Categories:

          Function(s) Process(es)

          anterograde axonal transport
          motor activity
          microtubule motor activity
          ATP binding
          microtubule binding
          ATPase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-55658555d6-7l2qm:80/100.66.75.127:80.
          git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.41.0-Offline