Search Thermo Fisher Scientific
- Contáctenos
- Orden Rápida
-
¿No tiene una cuenta? Crear una cuenta
Search Thermo Fisher Scientific
Gene Symbol: LOC100910678 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003749743.3 | XP_003749791.1 | 2 | 349 | ||
XM_006236476.2 | XP_006236538.1 | 1 | 501 |
Gene Symbol: Tma7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001126048.2 | NP_001119520.1 | 4 | 373 | ||
GenBank mRNA | BC157813.1 | - | 4 | 373 | ||
FQ221914.1 | - | 4 | 349 | |||
FQ222972.1 | - | 4 | 338 |