Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other BCAR1 SIRNA Assays ›
Gene Symbol:
BCAR1
Gene Name:
BCAR1, Cas family scaffolding protein
Gene Aliases:
CAS, CAS1, CASS1, CRKAS, P130Cas
Chromosome Location:
Chr.16: 75229030 - 75268053 on Build GRCh38
UniGene ID:
Hs.479747
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s18371
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
392.00
Your Price
Online offer:
380.65
380.65
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: BCAR1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001170714.1
NP_001164185.1
6
2419
NM_001170715.1
NP_001164186.1
5
2124
NM_001170716.1
NP_001164187.1
5
2081
NM_001170717.1
NP_001164188.1
5
2197
NM_001170718.1
NP_001164189.1
5
2168
NM_001170719.1
NP_001164190.1
5
2258
NM_001170720.1
NP_001164191.1
6
1829
NM_001170721.1
NP_001164192.1
4
2356
NM_014567.3
NP_055382.2
5
2143
XM_005256258.4
XP_005256315.1
5
2343
XM_005256259.4
XP_005256316.1
5
5255
XM_005256260.4
XP_005256317.1
5
2264
XM_011523475.2
XP_011521777.1
5
2292
XM_017023897.1
XP_016879386.1
5
2850
XM_017023898.1
XP_016879387.1
5
2850
XM_017023899.1
XP_016879388.1
3
2571
GenBank mRNA
AB040024.1
-
5
2138
AF218451.1
-
5
2121
AJ242987.2
-
5
2118
AK026121.1
-
1
627
AK027608.1
-
5
2168
AK074958.1
-
4
1553
AK124526.1
-
5
3563
AK124815.1
-
5
2127
AK124844.1
-
4
1934
AK125719.1
-
1
1602
AK293808.1
-
6
2419
AK294513.1
-
5
2174
AK295809.1
-
5
2053
AK302617.1
-
6
1829
AK314722.1
-
5
2143
AY545071.1
-
4
2051
BC014402.1
-
1
785
BC062556.1
-
4
1998
6427808
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
regulation of cell growth
actin filament organization
cell adhesion
epidermal growth factor receptor signaling pathway
G-protein coupled receptor signaling pathway
integrin-mediated signaling pathway
cell proliferation
insulin receptor signaling pathway
positive regulation of endothelial cell migration
cell migration
positive regulation of cell migration
cellular response to hepatocyte growth factor stimulus
regulation of apoptotic process
platelet-derived growth factor receptor signaling pathway
vascular endothelial growth factor receptor signaling pathway
neurotrophin TRK receptor signaling pathway
hepatocyte growth factor receptor signaling pathway
antigen receptor-mediated signaling pathway
T cell receptor signaling pathway
B cell receptor signaling pathway
cell division
cell chemotaxis
signal transducer activity
protein binding
SH3 domain binding
protein kinase binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0