Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Instrument Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
Our Instagram
Our Facebook
Blog Behind the Bench
Customer Experience Center (CEC)
Ecommerce Exclusives
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other BCAR1 SIRNA Assays ›
Gene Symbol:
BCAR1
Gene Name:
BCAR1, Cas family scaffolding protein
Gene Aliases:
CAS, CAS1, CASS1, CRKAS, P130Cas
Chromosome Location:
Chr.16: 75229030 - 75268053 on Build GRCh38
UniGene ID:
Hs.479747
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s18372
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
5,366.00
Your Price
Online offer:
5366.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: BCAR1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001170714.1
NP_001164185.1
6
2409
NM_001170715.1
NP_001164186.1
5
2114
NM_001170716.1
NP_001164187.1
5
2071
NM_001170717.1
NP_001164188.1
5
2187
NM_001170718.1
NP_001164189.1
5
2158
NM_001170719.1
NP_001164190.1
5
2248
NM_001170720.1
NP_001164191.1
6
1819
NM_001170721.1
NP_001164192.1
4
2346
NM_014567.3
NP_055382.2
5
2133
XM_005256258.4
XP_005256315.1
5
2333
XM_005256259.4
XP_005256316.1
5
5245
XM_005256260.4
XP_005256317.1
5
2254
XM_011523475.2
XP_011521777.1
5
2282
XM_017023897.1
XP_016879386.1
5
2840
XM_017023898.1
XP_016879387.1
5
2840
XM_017023899.1
XP_016879388.1
3
2561
GenBank mRNA
AB040024.1
-
5
2128
AF218451.1
-
5
2111
AJ242987.2
-
5
2108
AK026121.1
-
1
617
AK027608.1
-
5
2158
AK074958.1
-
4
1543
AK124526.1
-
5
3553
AK124815.1
-
5
2117
AK124844.1
-
4
1924
AK125719.1
-
1
1592
AK293808.1
-
6
2409
AK294513.1
-
5
2164
AK295809.1
-
5
2043
AK302617.1
-
6
1819
AK314722.1
-
5
2133
AY545071.1
-
4
2041
BC014402.1
-
1
775
BC062556.1
-
4
1988
6427808
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
regulation of cell growth
actin filament organization
cell adhesion
epidermal growth factor receptor signaling pathway
G-protein coupled receptor signaling pathway
integrin-mediated signaling pathway
cell proliferation
insulin receptor signaling pathway
positive regulation of endothelial cell migration
cell migration
positive regulation of cell migration
cellular response to hepatocyte growth factor stimulus
regulation of apoptotic process
platelet-derived growth factor receptor signaling pathway
vascular endothelial growth factor receptor signaling pathway
neurotrophin TRK receptor signaling pathway
hepatocyte growth factor receptor signaling pathway
antigen receptor-mediated signaling pathway
T cell receptor signaling pathway
B cell receptor signaling pathway
cell division
cell chemotaxis
signal transducer activity
protein binding
SH3 domain binding
protein kinase binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0