Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other BCAR1 SIRNA Assays ›
Gene Symbol:
BCAR1
Gene Name:
BCAR1, Cas family scaffolding protein
Gene Aliases:
CAS, CAS1, CASS1, CRKAS, P130Cas
Chromosome Location:
Chr.16: 75229030 - 75268053 on Build GRCh38
UniGene ID:
Hs.479747
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s18373
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
5,366.00
Your Price
Online offer:
5366.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: BCAR1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001170714.1
NP_001164185.1
7
2450
NM_001170715.1
NP_001164186.1
6
2155
NM_001170716.1
NP_001164187.1
6
2112
NM_001170717.1
NP_001164188.1
6
2228
NM_001170718.1
NP_001164189.1
6
2199
NM_001170719.1
NP_001164190.1
6
2289
NM_001170720.1
NP_001164191.1
7
1860
NM_001170721.1
NP_001164192.1
5
2387
NM_014567.3
NP_055382.2
6
2174
XM_005256258.4
XP_005256315.1
6
2374
XM_005256259.4
XP_005256316.1
6
5286
XM_005256260.4
XP_005256317.1
6
2295
XM_011523475.2
XP_011521777.1
6
2323
XM_017023897.1
XP_016879386.1
6
2881
XM_017023899.1
XP_016879388.1
4
2602
GenBank mRNA
AB040024.1
-
6
2169
AF218451.1
-
6
2152
AJ242987.2
-
6
2149
AK026121.1
-
2
658
AK027608.1
-
6
2199
AK074958.1
-
5
1584
AK124526.1
-
6
3594
AK124815.1
-
6
2158
AK124844.1
-
5
1965
AK125719.1
-
2
1633
AK293808.1
-
7
2450
AK294513.1
-
6
2205
AK295809.1
-
6
2084
AK302617.1
-
7
1860
AK314722.1
-
6
2174
AY545071.1
-
5
2082
BC014402.1
-
2
816
BC062556.1
-
5
2029
6427808
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
regulation of cell growth
actin filament organization
cell adhesion
epidermal growth factor receptor signaling pathway
G-protein coupled receptor signaling pathway
integrin-mediated signaling pathway
cell proliferation
insulin receptor signaling pathway
positive regulation of endothelial cell migration
cell migration
positive regulation of cell migration
cellular response to hepatocyte growth factor stimulus
regulation of apoptotic process
platelet-derived growth factor receptor signaling pathway
vascular endothelial growth factor receptor signaling pathway
neurotrophin TRK receptor signaling pathway
hepatocyte growth factor receptor signaling pathway
antigen receptor-mediated signaling pathway
T cell receptor signaling pathway
B cell receptor signaling pathway
cell division
cell chemotaxis
signal transducer activity
protein binding
SH3 domain binding
protein kinase binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0