Search
Search

Rad51ap1| Gene Symbol: Rad51ap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001079711.1 | NP_001073179.1 | 5 | 537 | ||
| XM_006237502.2 | XP_006237564.1 | 5 | 517 | |||
| XM_006237504.2 | XP_006237566.1 | 5 | 531 | |||
| GenBank mRNA | BC128747.1 | - | 5 | 537 | 7099220 | |